This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM193A SIRNA Assays ›
Gene Symbol:
FAM193A
Gene Name:
family with sequence similarity 193 member A
Gene Aliases:
C4orf8, RES4-22
Chromosome Location:
Chr.4: 2596288 - 2732575 on Build GRCh38
UniGene ID:
Hs.652364
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s16362

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM193A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256666.1 NP_001243595.1 16 2833
NM_001256667.1 NP_001243596.1 17 2899
NM_001256668.1 NP_001243597.1 16 2833
NM_003704.3 NP_003695.3 16 2833
NR_046335.1 16 2786
NR_046336.1 16 2846
XM_006713930.2 XP_006713993.2 17 2896
XM_006713932.2 XP_006713995.2 18 2968
XM_011513590.1 XP_011511892.1 19 3046
XM_011513593.2 XP_011511895.1 16 2767
XM_017008788.1 XP_016864277.1 18 2889
GenBank mRNA AB000459.1 - 16 2833
AB000460.1 - 16 2833
AB000461.1 - 16 2786
AF040966.1 - 12 2521
AK303732.1 - 12 2405
AK307430.1 - 8 1561
BC136646.1 - 16 2803 9020636
BC144337.1 - 16 2813
BC144339.1 - 16 2803
BC144340.1 - 17 2869 9052860

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0