This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OTUB2 SIRNA Assays ›
Gene Symbol:
Otub2
Gene Name:
OTU deubiquitinase, ubiquitin aldehyde binding 2
Gene Aliases:
-
Chromosome Location:
Chr.6: 127282444 - 127301086 on Build Rnor_6.0
UniGene ID:
Rn.18862
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s163193

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Otub2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108053.1 NP_001101523.1 2 213
XM_006240477.2 XP_006240539.1 3 553
GenBank mRNA BC168869.1 - 2 130 7104604
FQ211938.1 - 2 199

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_023112, AK025569, CR457345, AY177201, BC068058, and others.
This siRNA also targets the following transcripts in Mouse: NM_001177841, NM_026580, AK158758, AK076336, AK006469, and others.

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
protein deubiquitination
protein K11-linked deubiquitination
protein K63-linked deubiquitination
protein K48-linked deubiquitination
thiol-dependent ubiquitin-specific protease activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0