This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DFNB31 SIRNA Assays ›
Gene Symbol:
Dfnb31
Gene Name:
deafness, autosomal recessive 31
Gene Aliases:
Cip98, Whrn
Chromosome Location:
Chr.5: 79235541 - 79317206 on Build Rnor_6.0
UniGene ID:
Rn.231801
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s162329

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dfnb31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181088.1 NP_851602.1 2 653
GenBank mRNA AY227205.1 - 2 653

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
sensory perception of sound
positive regulation of gene expression
sensory perception of light stimulus
inner ear receptor stereocilium organization
protein domain specific binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header