This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYP4F2 SIRNA Assays ›
Gene Symbol:
CYP4F2
Gene Name:
cytochrome P450 family 4 subfamily F member 2
Gene Aliases:
CPF2
Chromosome Location:
Chr.19: 15878024 - 15898120 on Build GRCh38
UniGene ID:
Hs.558423
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s16215

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CYP4F2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082.4 NP_001073.3 6 657
GenBank mRNA AK296200.1 - 4 504
AK300961.1 - 5 463
BC067437.1 - 6 598 7002207
BC067438.1 - 6 598
BC067439.1 - 6 598 7002212
BC067440.1 - 6 598 7002214
D26480.1 - 6 602
U02388.2 - 6 620

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
long-chain fatty acid metabolic process
renal water homeostasis
pressure natriuresis
icosanoid metabolic process
leukotriene metabolic process
blood coagulation
regulation of blood pressure
drug metabolic process
arachidonic acid metabolic process
epoxygenase P450 pathway
negative regulation of icosanoid secretion
positive regulation of icosanoid secretion
leukotriene B4 catabolic process
vitamin E metabolic process
menaquinone catabolic process
phylloquinone catabolic process
vitamin K catabolic process
sodium ion homeostasis
oxidation-reduction process
omega-hydroxylase P450 pathway
monooxygenase activity
iron ion binding
protein binding
arachidonic acid epoxygenase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
alkane 1-monooxygenase activity
heme binding
leukotriene-B4 20-monooxygenase activity
arachidonic acid omega-hydroxylase activity
alpha-tocopherol omega-hydroxylase activity
tocotrienol omega-hydroxylase activity
20-hydroxy-leukotriene B4 omega oxidase activity
20-aldehyde-leukotriene B4 20-monooxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0