This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PHC2 SIRNA Assays ›
Gene Symbol:
Phc2
Gene Name:
polyhomeotic homolog 2 (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.5: 146832568 - 146931444 on Build Rnor_6.0
UniGene ID:
Rn.2428
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s162137

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Phc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013169.2 NP_001013187.2 9 1748
XM_006238926.2 XP_006238988.1 10 1819
XM_006238930.2 XP_006238992.1 2 466
XM_008764139.1 XP_008762361.1 2 431
GenBank mRNA BC169102.1 - 2 410

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_198040, NM_004427, BC028396, BC029269, BC092492, and others.
This siRNA also targets the following transcripts in Mouse: NM_018774, NM_001195130, NM_001195083, BC071246, AB062362, and others.

Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
biological_process
molecular_function
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0