This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GSAP SIRNA Assays ›
Gene Symbol:
Gsap
Gene Name:
gamma-secretase activating protein
Gene Aliases:
Pion
Chromosome Location:
Chr.4: 10423090 - 10517888 on Build Rnor_6.0
UniGene ID:
Rn.17968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s161630

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gsap
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107845.1 NP_001101315.1 12 849
XM_006235935.2 XP_006235997.1 12 903
XM_006235936.2 XP_006235998.1 12 905
XM_008762648.1 XP_008760870.1 12 905
XM_008762649.1 XP_008760871.1 12 904

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of proteolysis
positive regulation of beta-amyloid formation
beta-amyloid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0