This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SULF2 SIRNA Assays ›
Gene Symbol:
Sulf2
Gene Name:
sulfatase 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 162791349 - 162872986 on Build Rnor_6.0
UniGene ID:
Rn.4228
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s161356

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sulf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034927.1 NP_001030099.1 13 2539
XM_006235595.2 XP_006235657.1 13 2606
XM_006235596.2 XP_006235658.1 13 2137
XM_008762506.1 XP_008760728.1 13 2187
GenBank mRNA AY742216.1 - 13 2539
BC091377.1 - 3 318
BC166410.1 - 13 2018 9026145

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

kidney development
chondrocyte development
glomerular filtration
positive regulation of vascular endothelial growth factor production
esophagus smooth muscle contraction
positive regulation of Wnt signaling pathway
heparan sulfate proteoglycan metabolic process
glomerular basement membrane development
glial cell-derived neurotrophic factor receptor signaling pathway
negative regulation of fibroblast growth factor receptor signaling pathway
embryonic skeletal system development
cartilage development
bone development
innervation
arylsulfatase activity
calcium ion binding
N-acetylglucosamine-6-sulfatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0