This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TOMM34 SIRNA Assays ›
Gene Symbol:
Tomm34
Gene Name:
translocase of outer mitochondrial membrane 34
Gene Aliases:
-
Chromosome Location:
Chr.3: 160448294 - 160465143 on Build Rnor_6.0
UniGene ID:
Rn.16749
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s161334

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tomm34
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044244.2 NP_001037709.1 2 237
XM_006235585.2 XP_006235647.1 2 512
GenBank mRNA BC105768.1 - 2 238 7930355

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006809, BC014907, AK129538, AK222952, AB085681, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to mitochondrion
heat shock protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0