This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RPRD1B SIRNA Assays ›
Gene Symbol:
Rprd1b
Gene Name:
regulation of nuclear pre-mRNA domain containing 1B
Gene Aliases:
Crept, RGD1304782
Chromosome Location:
Chr.3: 154507029 - 154553312 on Build Rnor_6.0
UniGene ID:
Rn.4144
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s161309

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rprd1b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098727.1 NP_001092197.1 3 685
XM_006235412.1 XP_006235474.1 3 691
GenBank mRNA BC168699.1 - 3 690 8362231
DY310142.1 - 3 685
DY565371.1 - 3 461

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_021215, BC001696, BC033629, DQ372938, AK312468.
This siRNA also targets the following transcripts in Mouse: NM_027434, AK152220, AK147718, BC042447, AK152125, and others.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell proliferation
regulation of cell cycle process
positive regulation of transcription from RNA polymerase II promoter
dephosphorylation of RNA polymerase II C-terminal domain
RNA polymerase II core binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0