This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTLL9 SIRNA Assays ›
Gene Symbol:
Ttll9
Gene Name:
tubulin tyrosine ligase-like family, member 9
Gene Aliases:
-
Chromosome Location:
Chr.3: 148426057 - 148485370 on Build Rnor_6.0
UniGene ID:
Rn.138049
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s161265

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttll9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014051.1 NP_001014073.1 3 103
XM_006235276.2 XP_006235338.1 3 874
XM_008762331.1 XP_008760553.1 4 158
XM_008762332.1 XP_008760554.1 4 158
XM_008762333.1 XP_008760555.1 4 158
GenBank mRNA BC082105.1 - 3 103 7134735

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0