This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZC3H8 SIRNA Assays ›
Gene Symbol:
Zc3h8
Gene Name:
zinc finger CCCH type containing 8
Gene Aliases:
Zc3hdc8
Chromosome Location:
Chr.3: 121471836 - 121488528 on Build Rnor_6.0
UniGene ID:
Rn.137486
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s161157

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zc3h8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012090.1 NP_001012090.1 3 362
GenBank mRNA BC079122.1 - 3 362 7126707

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_032494, AK315329, BG773518, AF334161, BC032001.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
negative regulation of T cell differentiation in thymus
snRNA transcription from RNA polymerase II promoter
snRNA transcription from RNA polymerase III promoter
T cell homeostasis
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase III promoter
response to antibiotic
positive regulation of thymocyte apoptotic process
RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0