siRNA Details
Gene Symbol: Dusp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012089.1 | NP_001012089.1 | 2 | 574 | ||
GenBank mRNA | BC088205.1 | - | 2 | 574 | 7380627 |

Gene Symbol: Dusp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012089.1 | NP_001012089.1 | 2 | 574 | ||
GenBank mRNA | BC088205.1 | - | 2 | 574 | 7380627 |