This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IRS4 SIRNA Assays ›
Gene Symbol:
IRS4
Gene Name:
insulin receptor substrate 4
Gene Aliases:
IRS-4, PY160
Chromosome Location:
Chr.X: 108719949 - 108736584 on Build GRCh38
UniGene ID:
Hs.407141
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s16091

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IRS4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003604.2 NP_003595.1 1 3081
XM_005262220.2 XP_005262277.1 1 3288
XM_006724713.3 XP_006724776.1 1 3288
XM_011531061.1 XP_011529363.1 1 3288
GenBank mRNA AF007567.1 - 1 3126

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
insulin receptor signaling pathway
positive regulation of signal transduction
regulation of lipid metabolic process
signal transducer activity
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
phosphatidylinositol 3-kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0