This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CUL1 SIRNA Assays ›
Gene Symbol:
CUL1
Gene Name:
cullin 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 148698178 - 148801110 on Build GRCh38
UniGene ID:
Hs.146806
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s16054

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CUL1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003592.2 NP_003583.2 15, 16 2024
XM_005250060.4 XP_005250117.1 15, 16 2060
XM_011516629.2 XP_011514931.1 15, 16 2016
XM_011516630.2 XP_011514932.1 15, 16 1917
XM_011516631.2 XP_011514933.1 15, 16 2045
XM_011516632.2 XP_011514934.1 15, 16 1943
XM_017012723.1 XP_016868212.1 14, 15 1780
GenBank mRNA AF062536.1 - 14, 15 1798
AK096163.1 - 16, 17 1912
AK096505.1 - 8, 9 1681
AK296909.1 - 14, 15 1749
AK315023.1 - 15, 16 2200
BC125119.1 - 15, 16 1937 40118923
BC125120.1 - 15, 16 1937 40118927
BX537409.1 - 15, 16 1954
U58087.1 - 15, 16 1726

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
G2/M transition of mitotic cell cycle
protein polyubiquitination
stimulatory C-type lectin receptor signaling pathway
protein monoubiquitination
cell cycle arrest
cell proliferation
negative regulation of cell proliferation
organ morphogenesis
viral process
Wnt signaling pathway
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
T cell receptor signaling pathway
stress-activated MAPK cascade
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
intrinsic apoptotic signaling pathway
ubiquitin-protein transferase activity
protein binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0