This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARHGEF2 SIRNA Assays ›
Gene Symbol:
Arhgef2
Gene Name:
rho/rac guanine nucleotide exchange factor (GEF) 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 187964079 - 188022847 on Build Rnor_6.0
UniGene ID:
Rn.12255
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s160503

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arhgef2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012079.1 NP_001012079.1 10, 11 1303
XM_006232643.2 XP_006232705.1 15, 16 2081
XM_006232644.2 XP_006232706.1 16, 17 2198
XM_006232646.2 XP_006232708.1 14, 15 1658
XM_006232647.2 XP_006232709.1 13, 14 1502
XM_006232648.2 XP_006232710.1 13, 14 1453
XM_006232651.2 XP_006232713.1 10, 11 1591
XM_006232652.2 XP_006232714.1 10, 11 1410
XM_006232653.2 XP_006232715.1 10, 11 1298
XM_006232654.2 XP_006232716.1 10, 11 2107
XM_008761151.1 XP_008759373.1 16, 17 2193
XM_008761152.1 XP_008759374.1 11, 12 2181
GenBank mRNA BC090078.1 - 10, 11 1303 7125049
FQ212011.1 - 11, 12 1276

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
cell morphogenesis
actin filament organization
negative regulation of microtubule depolymerization
mitotic nuclear division
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
regulation of Rho protein signal transduction
intracellular signal transduction
innate immune response
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of neurogenesis
positive regulation of NF-kappaB transcription factor activity
cell division
cellular response to muramyl dipeptide
Rho guanyl-nucleotide exchange factor activity
microtubule binding
Rho GTPase binding
Rac guanyl-nucleotide exchange factor activity
metal ion binding
Rac GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0