siRNA Details
Gene Symbol: Cpne5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107616.1 | NP_001101086.1 | 4 | 297 | ||
GenBank mRNA | AM747282.1 | - | 4 | 199 |

Gene Symbol: Cpne5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107616.1 | NP_001101086.1 | 4 | 297 | ||
GenBank mRNA | AM747282.1 | - | 4 | 199 |