This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WBP1L SIRNA Assays ›
Gene Symbol:
Wbp1l
Gene Name:
WW domain binding protein 1-like
Gene Aliases:
Opal1, RGD1305793
Chromosome Location:
Chr.1: 266358728 - 266415296 on Build Rnor_6.0
UniGene ID:
Rn.105679
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s159786

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wbp1l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127484.1 NP_001120956.1 3 443
XM_006231508.2 XP_006231570.1 3 2438
XM_008760435.1 XP_008758657.1 3 1273
XM_008760436.1 XP_008758658.1 3 2550
GenBank mRNA BC166754.1 - 3 380 7929564
FQ217301.1 - 3 688

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0