This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANO1 SIRNA Assays ›
Gene Symbol:
Ano1
Gene Name:
anoctamin 1, calcium activated chloride channel
Gene Aliases:
Tmem16a
Chromosome Location:
Chr.1: 217754349 - 217931625 on Build Rnor_6.0
UniGene ID:
Rn.162509
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s159470

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ano1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107564.1 NP_001101034.1 17, 18 1992
XM_006230780.2 XP_006230842.1 17, 18 1994
XM_008760118.1 XP_008758340.1 18, 19 2073
XM_008760119.1 XP_008758341.1 17, 18 1912

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cation transport
chloride transport
regulation of membrane potential
trachea development
cation transmembrane transport
chloride transmembrane transport
calcium activated cation channel activity
intracellular calcium activated chloride channel activity
phospholipid scramblase activity
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0