This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDDC1 SIRNA Assays ›
Gene Symbol:
Pddc1 LOC100911365
Gene Name:
Parkinson disease 7 domain containing 1
Parkinson disease 7 domain-containing protein 1-like
Gene Aliases:
RGD1304836
-
Chromosome Location:
Chr.1: 214389047 - 214394421 on Build Rnor_6.0
Chromosome Location:
Chr.1: 213952109 - 213957542 on Build Rnor_6.0
UniGene ID:
Rn.99147
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s159446

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pddc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107563.2 NP_001101033.1 3 262
Gene Symbol: LOC100911365
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749006.3 XP_003749054.3 289

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header