This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FANK1 SIRNA Assays ›
Gene Symbol:
Fank1
Gene Name:
fibronectin type 3 and ankyrin repeat domains 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 205842458 - 205950360 on Build Rnor_6.0
UniGene ID:
Rn.214047
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s159412

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fank1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008347.1 NP_001008348.1 6, 7 618
XM_006230439.2 XP_006230501.1 6, 7 663
GenBank mRNA BC082095.1 - 6, 7 618 7134234

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025850, AK005694, AY819025, BC046278, AK017071.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0