This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDE8A SIRNA Assays ›
Gene Symbol:
Pde8a
Gene Name:
phosphodiesterase 8A
Gene Aliases:
RNPDE8A
Chromosome Location:
Chr.1: 143036019 - 143160395 on Build Rnor_6.0
UniGene ID:
Rn.24771
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s159125

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde8a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198767.1 NP_942062.1 3, 4 476
XM_006229438.2 XP_006229500.1 3, 4 663
GenBank mRNA AB092696.1 - 3, 4 476

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phosphorelay signal transduction system
biological_process
metabolic process
molecular_function
3',5'-cyclic-AMP phosphodiesterase activity
signal transducer activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0