This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCR6 SIRNA Assays ›
Gene Symbol:
Ccr6
Gene Name:
chemokine (C-C motif) receptor 6
Gene Aliases:
-
Chromosome Location:
Chr.1: 53063383 - 53087513 on Build Rnor_6.0
UniGene ID:
Rn.161767
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s158618

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccr6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013145.1 NP_001013163.1 2 659
XM_006227922.2 XP_006227984.1 3 769
XM_008758755.1 XP_008756977.1 2 536
XM_008758756.1 XP_008756978.1 2 2585
GenBank mRNA BC091197.1 - 2 659 7374663
EF547659.1 - 1 394
FQ225325.1 - 1 32
FQ225692.1 - 2 536
FQ226017.1 - 2 536
FQ227530.1 - 1 386

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001190337, NM_009835, NM_001190333, NM_001190335, NM_001190338, and others.

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
immune response
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
positive regulation of epithelial cell migration
chemokine-mediated signaling pathway
positive regulation of dendritic cell chemotaxis
molecular_function
C-C chemokine receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0