This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD226 SIRNA Assays ›
Gene Symbol:
Cd226
Gene Name:
CD226 molecule
Gene Aliases:
-
Chromosome Location:
Chr.18: 86299387 - 86394772 on Build Rnor_6.0
UniGene ID:
Rn.110297
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s157992

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd226
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107370.1 NP_001100840.1 5 946
XM_006255025.2 XP_006255087.1 6 1761
XM_006255028.2 XP_006255090.1 5 1397
XM_006255029.2 XP_006255091.1 6 1481
XM_006255031.2 XP_006255093.1 5 1117
XM_006255032.2 XP_006255094.1 6 1386

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytokine production
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
positive regulation of immunoglobulin mediated immune response
biological_process
positive regulation of mast cell activation
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of Fc receptor mediated stimulatory signaling pathway
molecular_function
integrin binding
protein kinase binding
cell adhesion molecule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0