This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other F12 SIRNA Assays ›
Gene Symbol:
F12
Gene Name:
coagulation factor XII (Hageman factor)
Gene Aliases:
HAF
Chromosome Location:
Chr.17: 9736577 - 9744420 on Build Rnor_6.0
UniGene ID:
Rn.53943
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s157719

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: F12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014006.1 NP_001014028.1 10 1338
GenBank mRNA BC088187.1 - 10 1338 7370526

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

plasma kallikrein-kinin cascade
Factor XII activation
proteolysis
blood coagulation
positive regulation of plasminogen activation
protein processing
protein autoprocessing
regulation of blood coagulation
positive regulation of blood coagulation
zymogen activation
fibrinolysis
response to misfolded protein
positive regulation of fibrinolysis
serine-type endopeptidase activity
peptidase activity
serine-type aminopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0