This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GOT1L1 SIRNA Assays ›
Gene Symbol:
Got1l1
Gene Name:
glutamic-oxaloacetic transaminase 1-like 1
Gene Aliases:
RGD1307172
Chromosome Location:
Chr.16: 69022413 - 69029926 on Build Rnor_6.0
UniGene ID:
Rn.36169
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s157579

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Got1l1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014003.1 NP_001014025.1 4 517
XM_006253291.2 XP_006253353.1 4 888
XM_006253292.2 XP_006253354.1 4 408
XM_006253294.2 XP_006253356.1 4 888
XM_008771326.1 XP_008769548.1 4 550
XM_008771327.1 XP_008769549.1 4 888
XM_008771328.1 XP_008769550.1 4 888
GenBank mRNA BC079187.1 - 4 517 7131193

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
biosynthetic process
transaminase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0