This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCTD6 SIRNA Assays ›
Gene Symbol:
Kctd6
Gene Name:
potassium channel tetramerization domain containing 6
Gene Aliases:
-
Chromosome Location:
Chr.15: 18484607 - 18493195 on Build Rnor_6.0
UniGene ID:
Rn.105101
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s157002

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kctd6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107253.1 NP_001100723.1 2 124
XM_006251763.2 XP_006251825.1 3 1759
XM_006251765.2 XP_006251827.1 2 2661
GenBank mRNA BC158799.1 - 1 70

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein homooligomerization
ankyrin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0