This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EIF4ENIF1 SIRNA Assays ›
Gene Symbol:
Eif4enif1
Gene Name:
eukaryotic translation initiation factor 4E nuclear import factor 1
Gene Aliases:
RGD1560908
Chromosome Location:
Chr.14: 83412886 - 83455694 on Build Rnor_6.0
UniGene ID:
Rn.105097
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s156831

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eif4enif1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107230.1 NP_001100700.1 18 2789
XM_006251290.2 XP_006251352.1 18 2921
XM_006251291.2 XP_006251353.1 18 2677
XM_008770296.1 XP_008768518.1 17 3384
GenBank mRNA FQ212399.1 - 18 2733

Back To Top

More Information


Gene Ontology Categories:

Process(es)

poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0