This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM193A SIRNA Assays ›
Gene Symbol:
Fam193a LOC103690085
Gene Name:
family with sequence similarity 193, member A
protein FAM193A-like
Gene Aliases:
RGD1309634
-
Chromosome Location:
Chr.14: 81515897 - 81638919 on Build Rnor_6.0
Chromosome Location:
Chr.14: 81897723 - 81909622 on Build Rnor_6.0
UniGene ID:
Rn.231962
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s156800

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam193a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_223536.8 XP_223536.5 19 3996
GenBank mRNA FQ225369.1 - 322
Gene Symbol: LOC103690085
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006251409.2 XP_006251471.1 6 2936

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm