This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TNIP2 SIRNA Assays ›
Gene Symbol:
Tnip2
Gene Name:
TNFAIP3 interacting protein 2
Gene Aliases:
-
Chromosome Location:
Chr.14: 81487665 - 81505018 on Build Rnor_6.0
UniGene ID:
Rn.17607
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s156798

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tnip2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024771.1 NP_001019942.1 3 713
XM_006251255.2 XP_006251317.1 3 1060
GenBank mRNA BC085912.1 - 3 644
BC097956.1 - 3 713 7461734

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_139064, AK160165, AJ304865, BC052083.

Gene Ontology Categories:

Function(s) Process(es)

I-kappaB kinase/NF-kappaB signaling
CD40 signaling pathway
toll-like receptor 2 signaling pathway
toll-like receptor 3 signaling pathway
toll-like receptor 9 signaling pathway
positive regulation of macrophage activation
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of transcription from RNA polymerase II promoter
protein stabilization
positive regulation of B cell activation
interleukin-1-mediated signaling pathway
cellular response to lipopolysaccharide
negative regulation of endothelial cell apoptotic process
protein kinase binding
polyubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0