This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VOM2R66 SIRNA Assays ›
Gene Symbol:
Vom2r66 LOC286980 Vom2r75
Gene Name:
vomeronasal 2 receptor, 66
putative pheromone receptor (Go-VN13B)
vomeronasal 2 receptor, 75
Gene Aliases:
RGD1566389
-
-
Chromosome Location:
Chr.14: 1388946 - 1415411 on Build Rnor_6.0
Chromosome Location:
-
Chromosome Location:
Chr.18: 11936 - 38488 on Build Rnor_6.0
UniGene ID:
-
Rn.10813
Rn.10813
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s156535

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vom2r66
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769953.1 XP_008768175.1 4 1129
Gene Symbol: LOC286980
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF016186.1 - 2 1012
Gene Symbol: Vom2r75
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173320.1 NP_775442.1 4 1518
XM_006254377.2 XP_006254439.1 4 1632
GenBank mRNA AF016185.1 - 4 1518

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0