This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP316 SIRNA Assays ›
Gene Symbol:
Zfp316
Gene Name:
zinc finger protein 316
Gene Aliases:
-
Chromosome Location:
Chr.12: 13346429 - 13367374 on Build Rnor_6.0
UniGene ID:
Rn.40708
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s155887

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp316
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107121.1 NP_001100591.1 5 737
XM_008768963.1 XP_008767185.1 5 3159
XM_008768964.1 XP_008767186.1 6 2810
XM_008768965.1 XP_008767187.1 5 2693
XM_008768966.1 XP_008767188.1 6 2750
XM_008768967.1 XP_008767189.1 5 2281
XM_008768968.1 XP_008767190.1 5 725

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
nucleic acid binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0