siRNA Details
Gene Symbol: Timmdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007676.1 | NP_001007677.1 | 5 | 635 | ||
GenBank mRNA | BC079140.1 | - | 5 | 635 | 7127856 | |
FQ215140.1 | - | 5 | 662 |

Gene Symbol: Timmdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007676.1 | NP_001007677.1 | 5 | 635 | ||
GenBank mRNA | BC079140.1 | - | 5 | 635 | 7127856 | |
FQ215140.1 | - | 5 | 662 |