This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IQCB1 SIRNA Assays ›
Gene Symbol:
Iqcb1
Gene Name:
IQ motif containing B1
Gene Aliases:
-
Chromosome Location:
Chr.11: 66824059 - 66878585 on Build Rnor_6.0
UniGene ID:
Rn.107714
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s155645

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Iqcb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107092.1 NP_001100562.1 14 1585
XM_006248395.2 XP_006248457.1 14 1651
XM_006248396.2 XP_006248458.1 14 1656
XM_006248397.2 XP_006248459.1 14 1619
XM_008768730.1 XP_008766952.1 14 1624

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cilium assembly
photoreceptor cell maintenance
maintenance of organ identity
calmodulin binding
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header