siRNA Details
Gene Symbol: Fn3krp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107077.2 | NP_001100547.2 | 1, 2 | 190 | ||
GenBank mRNA | BC166839.1 | - | 155 | 9027722 |

Gene Symbol: Fn3krp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107077.2 | NP_001100547.2 | 1, 2 | 190 | ||
GenBank mRNA | BC166839.1 | - | 155 | 9027722 |