This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB11FIP4 SIRNA Assays ›
Gene Symbol:
Rab11fip4
Gene Name:
RAB11 family interacting protein 4 (class II)
Gene Aliases:
RGD1305063
Chromosome Location:
Chr.10: 66942346 - 67051672 on Build Rnor_6.0
UniGene ID:
Rn.224682
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s155073

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab11fip4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107023.1 NP_001100493.1 14, 15 1836
XM_006246903.2 XP_006246965.1 14, 15 1819
XM_006246904.2 XP_006246966.1 12, 13 1781
XM_008768017.1 XP_008766239.1 14, 15 1713

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175543, AK046734, AK147344, AK147219, AK122559.

Gene Ontology Categories:

Process(es)

calcium ion binding
Rab GTPase binding
ADP-ribosylation factor binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0