siRNA Details
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127533.1 | NP_001121005.1 | 4 | 735 | ||
XM_006246250.2 | XP_006246312.1 | 4 | 778 | |||
GenBank mRNA | BC166514.1 | - | 4 | 717 | 7130556 |

More Information
Other Species Targeted

Gene Ontology Categories:
