This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARPC2 SIRNA Assays ›
Gene Symbol:
Arpc2
Gene Name:
actin related protein 2/3 complex, subunit 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 81518181 - 81548871 on Build Rnor_6.0
UniGene ID:
Rn.93317
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s154016

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arpc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106919.1 NP_001100389.1 4, 5 340
XM_006245165.1 XP_006245227.1 3, 4 478
XM_006245166.1 XP_006245228.1 4, 5 295
GenBank mRNA FQ214133.1 - 4, 5 300
FQ231947.1 - 1, 2 93

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of lamellipodium assembly
positive regulation of actin filament polymerization
Arp2/3 complex-mediated actin nucleation
positive regulation of substrate adhesion-dependent cell spreading
structural constituent of cytoskeleton
kinesin binding
AP-1 adaptor complex binding
actin filament binding
Arp2/3 complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0