This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100360552 SIRNA Assays ›
Gene Symbol:
LOC100360552
Gene Name:
rCG22430-like
Gene Aliases:
-
Chromosome Location:
Chr.9: 66305731 - 66308550 on Build Rnor_6.0
UniGene ID:
Rn.214809
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s153965

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100360552
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271185.1 NP_001258114.1 1 48

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008057, BC049781, U43320.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
G-protein coupled receptor signaling pathway
negative regulation of cell-substrate adhesion
skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
stem cell population maintenance
T cell differentiation in thymus
substrate adhesion-dependent cell spreading
regulation of catenin import into nucleus
non-canonical Wnt signaling pathway
non-canonical Wnt signaling pathway via JNK cascade
positive regulation of phosphorylation
negative regulation of ectodermal cell fate specification
positive regulation of transcription, DNA-templated
somatic stem cell division
positive regulation of epithelial cell proliferation involved in wound healing
canonical Wnt signaling pathway
mesenchymal to epithelial transition
G-protein coupled receptor activity
frizzled binding
Wnt-protein binding
PDZ domain binding
Wnt-activated receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0