This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCTD18 SIRNA Assays ›
Gene Symbol:
Kctd18
Gene Name:
potassium channel tetramerization domain containing 18
Gene Aliases:
RGD1564820
Chromosome Location:
Chr.9: 64849547 - 64916652 on Build Rnor_6.0
UniGene ID:
Rn.12331
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s153957

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kctd18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106914.1 NP_001100384.1 6 4035
XM_006244965.2 XP_006245027.1 4 2084
XM_006244966.2 XP_006245028.1 4 636
XM_006244967.2 XP_006245029.1 4 1148
XM_008767084.1 XP_008765306.1 3 2830
XM_008767085.1 XP_008765307.1 3 2948
XM_008767086.1 XP_008765308.1 4 641
XM_008767087.1 XP_008765309.1 4 570
GenBank mRNA FQ209862.1 - 5 1181
FQ228496.1 - 5 1204

Back To Top

More Information


Gene Ontology Categories:

Function(s)

protein homooligomerization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0