This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIM42 SIRNA Assays ›
Gene Symbol:
Trim42
Gene Name:
tripartite motif-containing 42
Gene Aliases:
-
Chromosome Location:
Chr.8: 105205040 - 105226580 on Build Rnor_6.0
UniGene ID:
Rn.109830
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s153663

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trim42
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013955.1 NP_001013977.1 4, 5 2233
XM_008766513.1 XP_008764735.1 4, 5 2278
GenBank mRNA BC087151.1 - 4, 5 2233 7131982

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_152616, BC130472, AK097416, BC130474, BX093220, and others.
This siRNA also targets the following transcripts in Mouse: NM_030219, AK019654.

Gene Ontology Categories:

Process(es)

zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0