siRNA Details
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047871.1 | NP_001041336.1 | 5 | 697 | ||
GenBank mRNA | BC161810.1 | - | 5 | 631 | 9023096 | |
DQ480751.1 | - | 5 | 697 |

Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047871.1 | NP_001041336.1 | 5 | 697 | ||
GenBank mRNA | BC161810.1 | - | 5 | 631 | 9023096 | |
DQ480751.1 | - | 5 | 697 |