This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BBS4 SIRNA Assays ›
Gene Symbol:
Bbs4
Gene Name:
Bardet-Biedl syndrome 4
Gene Aliases:
-
Chromosome Location:
Chr.8: 64121392 - 64154396 on Build Rnor_6.0
UniGene ID:
Rn.25002
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s153312

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bbs4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106826.1 NP_001100296.1 8 535
XM_008766224.1 XP_008764446.1 7 867
XM_008766225.1 XP_008764447.1 3 678
XM_008766226.1 XP_008764448.1 2 334

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
mitotic cytokinesis
neural tube closure
retina homeostasis
negative regulation of systemic arterial blood pressure
spermatid development
sensory perception of smell
protein localization
biological_process
metabolic process
negative regulation of gene expression
dendrite development
regulation of lipid metabolic process
ventricular system development
striatum development
hippocampus development
cerebral cortex development
adult behavior
negative regulation of actin filament polymerization
regulation of cytokinesis
leptin-mediated signaling pathway
protein localization to organelle
microtubule anchoring at centrosome
nonmotile primary cilium assembly
social behavior
photoreceptor cell outer segment organization
negative regulation of appetite by leptin-mediated signaling pathway
positive regulation of multicellular organism growth
cilium assembly
response to leptin
fat cell differentiation
photoreceptor cell maintenance
positive regulation of cilium assembly
retinal rod cell development
brain morphogenesis
centrosome organization
maintenance of protein location in nucleus
regulation of stress fiber assembly
cilium morphogenesis
regulation of cilium beat frequency involved in ciliary motility
face development
fat pad development
protein localization to cilium
protein localization to centrosome
regulation of nonmotile primary cilium assembly
protein localization to photoreceptor outer segment
RNA polymerase II repressing transcription factor binding
molecular_function
microtubule motor activity
dynactin binding
alpha-tubulin binding
beta-tubulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0