This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ABCG4 SIRNA Assays ›
Gene Symbol:
Abcg4
Gene Name:
ATP-binding cassette, subfamily G (WHITE), member 4
Gene Aliases:
-
Chromosome Location:
Chr.8: 48604915 - 48626214 on Build Rnor_6.0
UniGene ID:
Rn.29008
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s153220

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Abcg4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106816.1 NP_001100286.1 14 2036
XM_006242915.2 XP_006242977.1 14 7756
XM_006242916.2 XP_006242978.1 14 2114
GenBank mRNA BC089097.1 - 2 142

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
cholesterol efflux
ATP binding
ATPase activity
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0