This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM218 SIRNA Assays ›
Gene Symbol:
Tmem218
Gene Name:
transmembrane protein 218
Gene Aliases:
RGD1311364
Chromosome Location:
Chr.8: 39687281 - 39703455 on Build Rnor_6.0
UniGene ID:
Rn.105997
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s153033

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem218
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008325.1 NP_001008326.1 2, 3 157
XM_006242802.2 XP_006242864.1 2, 3 272
XM_008766050.1 XP_008764272.1 2, 3 308
GenBank mRNA BC085342.1 - 2, 3 157 7303171
FQ214300.1 - 2, 3 177

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025464, BC034160, AK003010, AK161115, AK008902.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0