This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF202 SIRNA Assays ›
Gene Symbol:
ZNF202
Gene Name:
zinc finger protein 202
Gene Aliases:
ZKSCAN10, ZSCAN42
Chromosome Location:
Chr.11: 123723927 - 123741705 on Build GRCh38
UniGene ID:
Hs.112556
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s15272

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF202
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301779.1 NP_001288708.1 8 1468
NM_001301780.1 NP_001288709.1 7 1324
NM_001301819.1 NP_001288748.1 6 1185
NM_003455.3 NP_003446.2 9 1520
XM_005271659.1 XP_005271716.1 9 1552
XM_005271660.1 XP_005271717.1 9 1561
XM_005271661.1 XP_005271718.1 8 1432
XM_005271664.1 XP_005271721.1 7 1257
XM_006718901.2 XP_006718964.1 10 1625
XM_011542972.1 XP_011541274.1 8 1721
XM_011542973.1 XP_011541275.1 9 1831
XM_011542975.1 XP_011541277.1 9 1994
XM_011542976.1 XP_011541278.1 8 1717
XM_017018268.1 XP_016873757.1 10 1692
GenBank mRNA AF027218.1 - 6 1098
AF027219.1 - 6 1102
AF155101.1 - 6 790
AK023751.1 - 7 1258
AK226047.1 - 8 1467
AL162031.1 - 6 1052
BC013382.2 - 9 1492 3508386

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
lipid metabolic process
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0