This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDRG1 SIRNA Assays ›
Gene Symbol:
Ndrg1
Gene Name:
N-myc downstream regulated 1
Gene Aliases:
Ndr1
Chromosome Location:
Chr.7: 107734323 - 107775701 on Build Rnor_6.0
UniGene ID:
Rn.153992
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s152526

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndrg1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011991.1 NP_001011991.1 13 983
XM_006241708.2 XP_006241770.1 13 1255
GenBank mRNA AY500369.1 - 13 945
BC081898.1 - 13 983 7110455
FQ212154.1 - 13 995

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA damage response, signal transduction by p53 class mediator
peripheral nervous system myelin maintenance
mast cell activation
cellular response to hypoxia
microtubule binding
Rab GTPase binding
gamma-tubulin binding
cadherin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0