This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WDR35L SIRNA Assays ›
Gene Symbol:
Wdr35l
Gene Name:
WD repeat domain 35-like
Gene Aliases:
RGD1564116, Wdr35
Chromosome Location:
Chr.6: 34094290 - 34152048 on Build Rnor_6.0
UniGene ID:
Rn.14574
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s151793

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wdr35l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099340.1 NP_001092810.1 22 2709
XM_006239885.2 XP_006239947.1 23 2758
GenBank mRNA EF613262.1 - 22 2709

Back To Top

More Information


Gene Ontology Categories:

Function(s)

intraciliary retrograde transport
cilium assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0