This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF223 SIRNA Assays ›
Gene Symbol:
ZNF223 ZNF222
Gene Name:
zinc finger protein 223
zinc finger protein 222
Gene Aliases:
-
-
Chromosome Location:
Chr.19: 44051372 - 44069902 on Build GRCh38
Chromosome Location:
Chr.19: 44025324 - 44033110 on Build GRCh38
UniGene ID:
Hs.720813
Hs.279840
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s15147

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF223
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF187989.1 - 5 1259
Gene Symbol: ZNF222
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129996.1 NP_001123468.1 4 1143
NM_013360.2 NP_037492.2 4 1111
XM_005259208.3 XP_005259265.1 5 1207
XM_005259209.3 XP_005259266.1 3 1064
GenBank mRNA AF187988.1 - 4 1090
AK095196.1 - 6 1303
BC030261.1 - 4 1085 5210539

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0