This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM59 SIRNA Assays ›
Gene Symbol:
Tmem59
Gene Name:
transmembrane protein 59
Gene Aliases:
RGD1310313
Chromosome Location:
Chr.5: 126783061 - 126804177 on Build Rnor_6.0
UniGene ID:
Rn.105907
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s151276

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem59
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139465.2 NP_001132937.1 8 899
GenBank mRNA BF287725.1 - 4 248
CV126271.1 - 2 76
FQ214069.1 - 8 897
FQ226828.1 - 7 619
FQ231829.1 - 8 885
FQ232106.1 - 8 904

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_004872, AF047439, BC095464, AX775909, BC016374, and others.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
positive regulation of autophagy
negative regulation of protein processing
negative regulation of establishment of protein localization to plasma membrane
negative regulation of protein glycosylation in Golgi
endopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0