This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF79 SIRNA Assays ›
Gene Symbol:
ZNF79
Gene Name:
zinc finger protein 79
Gene Aliases:
pT7
Chromosome Location:
Chr.9: 127424374 - 127446289 on Build GRCh38
UniGene ID:
Hs.522399
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s15126

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF79
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286696.1 NP_001273625.1 5 978
NM_001286697.1 NP_001273626.1 6 980
NM_001286698.1 NP_001273627.1 3 755
NM_001322260.1 NP_001309189.1 6 1109
NM_007135.2 NP_009066.2 5 871
XM_006717279.3 XP_006717342.1 6 1079
GenBank mRNA AK054606.1 - 5 863
AK302284.1 - 5 920
AK316233.1 - 6 916
BC062309.1 - 5 826 3944657

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
RNA polymerase II regulatory region sequence-specific DNA binding
molecular_function
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0